Key Documents
504335
Supelco
SPE Tube Adapter
PTFE (with female luer port), for use with 6 mL glass SPE tubes, pk of 24
Select a Size
24 EA
$536.00
$536.00
Available to ship onApril 16, 2025Details
Request a Bulk OrderRequest a Sample
Select a Size
Change View
24 EA
$536.00
About This Item
UNSPSC Code:
12352200
$536.00
Available to ship onApril 16, 2025Details
Request a Bulk OrderRequest a Sample
material
PTFE (with female luer port)
Quality Level
packaging
pk of 24
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 6 mL glass SPE tubes
General description
Tube adapters serve many purposes. They can be used to stack one SPE tube on top of another to provide different selectivities. A larger empty syringe barrel can be stacked on top of a smaller SPE tube to act as a larger load reservoir. Or, they can serve as an adapter for positive pressure methods (e.g. from a syringe or air/ N2 line).
Choose from one of the most recent versions:
Certificates of Analysis (COA)