Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21018-U

Supelco

Cartridge Adapter for S10, H10, H30 Cartridges

pkg of 10 ea

Synonym(s):

SPE cartridge tube Adapter

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$37.30

$37.30


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
10 EA
$37.30

About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

$37.30


Available to ship onApril 07, 2025Details


Request a Bulk Order

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with 1, 3 & 6 mL SPE cartridges

General description

Cartridge Adapter for S10, H10, H30 Cartridges is an accessory for use with LpDNPH cartridges, which are air sampling devices designed for sampling carbonyls (e.g., formaldehyde) in ambient, indoor, workplace, and source atmospheres.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service