Key Documents
502367
Micro-Metering Valve (Low Pressure)
1/4-28 flangeless fittings
Select a Size
1 EA
$123.00
$123.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$123.00
About This Item
UNSPSC Code:
41115854
eCl@ss:
32060101
$123.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PEEK
description
micro-metering low pressure
parameter
800 psi max. pressure
fitting
1/4-28 flangeless fittings
General description
Micro-Metering Valve is designed for the control of fluid flow rates. These needle valves are ideally used with peristaltic pumps, pump fluid-transfer, mass spectrometry and fraction collection.
Application
Micro-Metering valve (low pressure) can be used as a component of supercritical fluid extraction apparatus employing supercritical carbon dioxide, which may be used in the extraction and refining of soya-bean oil from full fat soya bean flakes[1] and essential oil from the roots of Vetiveria zizanioides.[2]
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Response surface method applied to supercritical carbon dioxide extraction of Vetiveria zizanioides essential oil.
Danh T L, et al.
Chemical Engineering Journal, 155(3), 617-626 (2009)
Characterization of soybean oil extracted by supercritical carbon dioxide and hexane.
Friedrich P J and List R G.
Journal of Agricultural and Food Chemistry, 30(1), 192-193 (1982)