Key Documents
22117
Fine Metering Valve
stainless steel, 1/8 in. straight arms
Synonym(s):
Swagelok® SS2,SS METERING VALVE 1/8S EA
Select a Size
1 EA
$553.00
$553.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$553.00
About This Item
UNSPSC Code:
41115717
eCl@ss:
32035302
$553.00
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel
fitting
1/8 in. straight arms
General description
Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Critical micelle concentrations for alkyltrimethylammonium bromides in water from 25 to 160 C.
Fennell ED
Journal of Solution Chemistry, 13 (2), 87-101 (1984)