Skip to Content
MilliporeSigma
All Photos(1)

Documents

46944-U

Supelco

(−) Quinic acid

analytical standard

Synonym(s):

D-(−)-Quinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O6
CAS Number:
Molecular Weight:
192.17
Beilstein:
2212412
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

O[C@@H]1C[C@@](O)(C[C@@H](O)[C@H]1O)C(O)=O

InChI

1S/C7H12O6/c8-3-1-7(13,6(11)12)2-4(9)5(3)10/h3-5,8-10,13H,1-2H2,(H,11,12)/t3-,4-,5-,7+/m1/s1

InChI key

AAWZDTNXLSGCEK-WYWMIBKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu Yang et al.
Journal of AOAC International, 95(4), 1053-1058 (2012-09-14)
Because almost every traditional Chinese medicine (TCM) is a multicomponent system, QC of TCMs always involves various difficulties. As a current popular quality assessment approach, focusing on qualitative and quantitative analysis of certain compounds contained in herbal medicine has been
Shao-xia Wang et al.
Chinese journal of integrative medicine, 18(6), 437-444 (2012-07-24)
To investigate the effects of caffeic acid ester fraction (Caf) from Erigeron breviscapus, mainly composed of dicaffeoylquinic acids (diCQAs), on microglial activation in vitro and focal cerebral ischemia in vivo. The production of nitric oxide (NO), tumor necrosis factor α
Yuan-Yuan Xie et al.
Journal of agricultural and food chemistry, 60(51), 12574-12583 (2012-12-12)
A multicomponent quantification fingerprint based on HPLC coupled with diode array detection and electrospray ionization tandem mass spectrometry (HPLC-DAD-ESI/MS) technique has been established for the analysis of phenolic compounds in 12 samples originated from 5 different cultivars of Chrysanthemum morifolium
Joo Youn Kim et al.
Archives of pharmacal research, 35(6), 1115-1122 (2012-08-09)
Ilex latifolia (Aquifoliaceae), one of the primary components of "Ku-ding-cha", has been used in Chinese folk medicine to treat headaches and various inflammatory diseases. A previous study demonstrated that the ethanol extract of I. latifolia could protect against ischemic apoptotic
Yong-Jin Yang et al.
Food chemistry, 138(2-3), 2057-2063 (2013-02-16)
Five new quinic acid derivatives and two known 3-O-feruloylquinic acids were isolated from the roots of Scorzonera divaricata Turcz. The new compounds were elucidated as (-)-1,4-di-O-feruloyl-3-O-dihydrocaffeoylquinic acid, (-)-1-O-feruloyl-4-O- dihydrocaffeoylquinic acid, (-)-3,5-di-O-feruloylquinic acid, (-)-1-O-feruloyl-3-O-dihydro- caffeoylquinic acid, and (-)-1-O-feruloyl-5-O-dihydrocaffeoylquinic acid based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service