MilliporeSigma
  • Caffeic acid ester fraction from Erigeron breviscapus inhibits microglial activation and provides neuroprotection.
Caffeic acid ester fraction from Erigeron breviscapus inhibits microglial activation and provides neuroprotection.
Chinese journal of integrative medicine (2012-07-24)
Shao-xia Wang, Hong Guo, Li-min Hu, Ya-nan Liu, Yue-fei Wang, Li-yuan Kang, Xiu-mei Gao
ABSTRACT
To investigate the effects of caffeic acid ester fraction (Caf) from Erigeron breviscapus, mainly composed of dicaffeoylquinic acids (diCQAs), on microglial activation in vitro and focal cerebral ischemia in vivo. The production of nitric oxide (NO), tumor necrosis factor α (TNF-α), and interleukin-1β (IL-1β) induced by lipopolysaccharide (LPS) treatment in rat primary cultured microglia were measured by Griess reaction or enzyme-linked immunosorbent assay. Cell viability of cortical neurons was measured using AlamarBlue reagent. The behavioral tests and the infarct area of brain were used to evaluate the damage to central nervous system in rat middle cerebral artery occlusion (MCAO) model of cerebral ischemia. Real time polymerase chain reaction was used to determine the expression of inducible nitric oxide synthase (iNOS), TNF-α and IL-1β mRNA in ischemic cerebral tissues. Caf inhibited the production of NO, TNF-α and IL-1β induced by LPS treatment in primary microglia in a dose-dependent manner. Exposure of cortical neurons to conditioned medium from Caf-treated microglia increased neuronal cell viability (P<0.01) compared with conditioned medium from LPS-treated alone. In MCAO rat model of cerebral ischemia, Caf could significantly improve neurobehavioural performance and reduce percentage infarct volume compared with the vehicle group (P<0.05). Caf could also significantly inhibit the up-regulation of iNOS, TNF-α, and IL-1β gene expressions in ischemic cerebral tissues. Caf could suppress microglial activation, which may be one mechanism of its neuroprotective effect against ischemia.
Sigma-Aldrich
D-(−)-Quinic acid, 98%