Skip to Content
MilliporeSigma
All Photos(1)

Documents

442700

Supelco

Pentadecane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH3
CAS Number:
Molecular Weight:
212.41
Beilstein:
1698194
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

7.4 (vs air)

vapor pressure

1 mmHg ( 91.6 °C)

description

This product has been discontinued. It has been replaced by product 76509.

expl. lim.

6.5 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.431 (lit.)

bp

270 °C (lit.)

mp

8-10 °C (lit.)

density

0.769 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCC

InChI

1S/C15H32/c1-3-5-7-9-11-13-15-14-12-10-8-6-4-2/h3-15H2,1-2H3

InChI key

YCOZIPAWZNQLMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthesis of phomactins: common intermediate phomactatriene and taxadiene.
Tetsuo Tokiwano et al.
Chemical communications (Cambridge, England), (11)(11), 1324-1325 (2004-05-22)
Keith D Schwartz et al.
Organic letters, 13(2), 248-251 (2010-12-18)
Conjugate reduction of an enone accompanied by in situ intramolecular aldol condensation was used to construct the tetrasubstituted cyclohexane nucleus of phomactins. Subsequent relay ring-closing metathesis completed the nine-membered ansa bridge of the diterpenoid framework.
Synthesis of homogeneous FePt nanoparticles using a nitrile ligand.
Virginie Monnier et al.
Small (Weinheim an der Bergstrasse, Germany), 4(8), 1139-1142 (2008-07-16)
From 2,2'-methylenedifuran to all stereomeric pentadecane-1,3,5,7,9,11,13,15-octols.
M E Schwenter et al.
The Journal of organic chemistry, 66(23), 7869-7872 (2001-11-10)
Heber J Chacon-Madrid et al.
Environmental science & technology, 46(20), 11179-11186 (2012-09-14)
We use a two-dimensional volatility basis set (2D-VBS) box model to simulate secondary organic aerosol (SOA) mass yields of linear oxygenated molecules: n-tridecanal, 2- and 7-tridecanone, 2- and 7-tridecanol, and n-pentadecane. A hybrid model with explicit, a priori treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service