Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442456

Supelco

Anthracene-d10

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D10
CAS Number:
Molecular Weight:
188.29
Beilstein:
2055675
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

340 °C (lit.)

mp

210-215 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+10

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c2c([2H])c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H]

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

MWPLVEDNUUSJAV-LHNTUAQVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anthracene-d10 has been used as internal standard in the analysis of pesticides in water samples, using gas chromatography coupled with mass spectrometry (GC-MS). It has also been used as an internal standard in the determination of polycyclic aromatic hydrocarbons in water samples using magnetic solid phase extraction technique and gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Monitoring of pesticides in drinking and related waters in NE Spain with a multiresidue SPE-GC?MS method including an estimation of the uncertainty of the analytical results
Quintana J, et al.
Journal of Chromatography A, 938, 3-13 (2001)
n-Octadecylphosphonic acid grafted mesoporous magnetic nanoparticle: Preparation, characterization, and application in magnetic solid-phase extraction
Ding J, et al.
Journal of Chromatography A, 1217, 7351-7358 (2010)
Tzu-Ling Chen et al.
Journal of chromatography. A, 1422, 340-344 (2015-11-01)
The rapid screening of trace amounts of the nitrogenous disinfection by-products, haloacetamides (HAcAms), in drinking and swimming pool water was performed by a simple and reliable procedure based on salt-assisted liquid-liquid extraction (SALLE) combined with injection-port silylation gas chromatography-mass spectrometry
Ying Li et al.
Environmental science and pollution research international, 22(23), 18446-18455 (2015-08-14)
The effects of ultrasonic irradiation, the chelating agent modified Fenton reaction, and a combination of ultrasound and the Fenton method in removing petroleum contaminants from a soil were studied. The results showed that the contaminant removal rate of the Fenton
Shinji Takeuchi et al.
The Science of the total environment, 491-492, 28-33 (2014-04-29)
Various plasticizers and flame retardants are contained in building materials and furniture produced for indoor environments. However, some of these material inclusions have been reported to cause endocrine-disrupting and mucosa-irritating effects. Because of the local climate, buildings in Sapporo are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service