MilliporeSigma
  • Rapid screening of haloacetamides in water using salt-assisted liquid-liquid extraction coupled injection-port silylation gas chromatography-mass spectrometry.
Rapid screening of haloacetamides in water using salt-assisted liquid-liquid extraction coupled injection-port silylation gas chromatography-mass spectrometry.
Journal of chromatography. A (2015-11-01)
Tzu-Ling Chen, Shin-Hwa Tzing, Wang-Hsien Ding
ABSTRACT
The rapid screening of trace amounts of the nitrogenous disinfection by-products, haloacetamides (HAcAms), in drinking and swimming pool water was performed by a simple and reliable procedure based on salt-assisted liquid-liquid extraction (SALLE) combined with injection-port silylation gas chromatography-mass spectrometry (IPS-GC-MS) method. The optimal SALLE conditions involved the injection of 4-mL of ethyl acetate into a 10-mL water sample (pH 7) containing 3-g of sodium sulfate. After vortex extraction for 1min and centrifugation, 10μL of the extract (mixed with 1μL of MTBSTFA) was directly determined by IPS-GC-MS. The limits of quantitation (LOQs) were determined to be 0.03-0.3μg/L. Precision, as indicated by relative standard deviations (RSDs), was less than 10% for both intra- and inter-day analysis. Accuracy, expressed as the mean extraction recovery, was between 76% and 94%. The SALLE plus IPS-GC-MS was successfully applied to quantitatively determine HAcAms from drinking and swimming pool water samples, and the total concentrations of the compounds ranged from 0.43 to 4.03μg/L.
Sigma-Aldrich
Anthracene-d10, 98 atom % D
Pricing and availability is not currently available.