Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33085

Supelco

Acetic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
Beilstein:
385737
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

for GC derivatization

vapor density

3.5 (vs air)

vapor pressure

10 mmHg ( 36 °C)
4 mmHg ( 20 °C)

autoignition temp.

629 °F

expl. lim.

10.3 %

packaging

pkg of 10 × 2 mL

refractive index

n20/D 1.390 (lit.)

bp

138-140 °C (lit.)

mp

−73 °C (lit.)

solubility

H2O: slightly soluble

density

1.08 g/mL (lit.)

SMILES string

CC(=O)OC(C)=O

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

General description

Acetic anhydride (CH3CO)2O, also known as ethanoric anhydride, is a colorless liquid with a strong pungent odor. It has a boiling point of 140°C (284°F). It is soluble in cold water, decomposes in hot water to form acetic acid, and is soluble in alcohol, chloroform and ether. It is a highly corrosive flammable liquid with harmful vapour. Exposure to acetic anhydride can occur via eye or skin contact, ingestion and inhalation.

Application

Suitable for the derivatization of acetaminophen, alcohols, amines, amino acids, arginyl peptides, aryl sulfate esters, carbamates, cerebrosides, cystine and cysteine (in peptides), desmethyldoxepin, 3,4-dihydroxyphenylglycol, hydroxamic acids, hydroxyamines, n-hydroxycarbamates, 4-methoxy and 3,4-dimethoxyphenethylamine, nucleosides bases and sugars, peptides, phenolalkylamines, phenylethylamines, phenylthiohydantoin amino acid, serotonin, and tryptamines.

Other Notes

Reagent for acetyl, o-acetyl, N-acetyl-N, permethyl acetyl, aryl acetate, ß-carboline, methyl ester, acetyl or trifluoroacetyl phenylthihydantoins, n-trifluoroacetyl, bis-trimethylsilyl, trimethylsilyl polyamino alcohol, and O-trimethylsilyl (TMS).

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. What is the advantage of acylation compared to silylation?

    Acylation, an alternative to silylation, produces stable, volatile derviatives of phenols, alcohols, and amines for analysis by GC/FID.  Acylated compounds are more stable than their corresponding silylated compounds.  

  4. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  5. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Handbook of Corrosion Data.
Craig BD and Anderson DS.
Technology & Engineering, 108-109 (1994)
Safe Use of Chemicals: A Practical Guide.
Dikshith
Technology & Engineering, 44-44 (2008)
J M Mee et al.
Biomedical mass spectrometry, 4(3), 178-181 (1977-06-01)
A quantitative analysis of amino acids in microsamples of dried blood spots by chemical ionization mass spectrometry has been developed. Isotope ratio determination was used as the quantitating technique via multiple labelled internal standards. This procedure yields excellent precision and
Brain tryptamine in rats by a new gas chromatography-mas fragmentographic method.
J J Warsh et al.
Biochemical medicine, 18(1), 10-20 (1977-08-01)
David G Simmons et al.
BMC genomics, 9, 352-352 (2008-07-30)
The Prolactin (PRL) hormone gene family shows considerable variation among placental mammals. Whereas there is a single PRL gene in humans that is expressed by the pituitary, there are an additional 22 genes in mice including the placental lactogens (PL)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service