MilliporeSigma
  • Rapid and quantitative blood amino acid analysis by chemical ionization mass spectrometry.
Rapid and quantitative blood amino acid analysis by chemical ionization mass spectrometry.
Biomedical mass spectrometry (1977-06-01)
J M Mee, J Korth, B Halpern, L B James
PMID890078
ABSTRACT
A quantitative analysis of amino acids in microsamples of dried blood spots by chemical ionization mass spectrometry has been developed. Isotope ratio determination was used as the quantitating technique via multiple labelled internal standards. This procedure yields excellent precision and accuracy as demonstrated by the analysis of known amino acid mixtures and of phenylalanine in the blood from newborns.
Product Description
Supelco
Acetic anhydride, LiChropur, ≥99.0%