Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27759-U

domnick hunter® Zero Air Generator

Model 35, 110 volt, output flow, 3500 cc/min

Synonym(s):

Gas generator, Pure Air, Zero Air

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$12,540.00

$12,540.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$12,540.00

About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

$12,540.00


Please contact Customer Service for Availability

Request a Bulk Order

feature

output flow 3500 cc/min

manufacturer/tradename

Domnick Hunter UHP-35ZA

Looking for similar products? Visit Product Comparison Guide

General description

  • Low power usage
  • Easy to wall mount
  • CE approved

Nitrox Zero Air Generators employ a catalytic oxidation system to produce a continuous supply of air that is virtually free of hydrocarbons, including methane. After incoming compressed air is filtered to remove oil, water, and particles, it is passed over a heated platinum catalyst that converts methane and oxygen in the air to carbon dioxide and water. If the hydrocarbon content of the input air is less than 10 ppm (as methane), the residual hydrocarbon concentration (as methane) is less than 0.1 ppm. A high efficiency 0.01 micron filter downstream of the catalyst ensures further removal of particles from the product stream.

Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at [email protected] or phone 800-359-3041.

Legal Information

domnick hunter is a registered trademark of Parker Hannifin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service