Key Documents
27749-U
domnick hunter® NITROX UHP Hydrogen Generator
Model 40H, output flow, 0 ‑ 250 cc/min
Select a Size
1 EA
$19,610.00
$19,610.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$19,610.00
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
$19,610.00
Please contact Customer Service for Availability
Request a Bulk Order
feature
output flow 0 ‑ 250 cc/min
manufacturer/tradename
Domnick Hunter 40H
General description
domnick hunter hydrogen generators are a safe and economic alternative to high pressure gas cylinders. These generators deliver a continuous stream of 99.999% pure hydrogen, suitable for use with any laboratory instrument requiring a supply of ultra high purity hydrogen, and ideal for carrier gas and fuel gas applications.
domnick hunter hydrogen generators use a proprietary ion exchange membrane to produce ultra-high-purity hydrogen from laboratory grade deionized water, totally eliminating the need for caustic solution.
These generators are designed for total safety and highly reliable operation. Hydrogen is generated only on demand, at low pressure, and minimal volume is stored within the unit. The unique digital display gives system status and diagnostic capabilities not available on any other competitive hydrogen generator.
CE Approved.
Legal Information
NITROX is a trademark of domnick hunter group PLC
domnick hunter is a registered trademark of Parker Hannifin
Choose from one of the most recent versions:
Certificates of Analysis (COA)