Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24418

Syringes for Shimadzu AOC14, 17, 20 and 20i Autosamplers

volume 10 μL, 10R-S-0.47, needle size 26 ga, needle L 42 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$168.00

$168.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$168.00

About This Item

UNSPSC Code:
41122001

$168.00


Please contact Customer Service for Availability

Request a Bulk Order

material

cone tip stainless steel needle (point style 1)

needle size

26 ga

feature

color-coded dark orange patch on barrel for easy volume identification barrel

packaging

pkg of 1 ea

manufacturer/tradename

SGE 002897

needle L

42 mm

volume

10 μL

compatibility

for use with Shimadzu AOC14 and AOC17 GC Autosamplers

Looking for similar products? Visit Product Comparison Guide

General description

Autosampler allows introduction of samples from vials into LC or GC systems. It generally includes motorized sample syringes which help in drawing the samples from vial. The use of autosampler and syringe can remove human error and increase accuracy/precision of the injection.

Legal Information

Shimadzu is a trademark of Shimadzu Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service