Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22000-U

Swagelok® Nut

Swagelok® 402-1, brass, 1/4 in. Swagelok

Synonym(s):

Swagelok® 402-1,BRASS NUTS 1/4"PK20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

material

brass

packaging

pkg of 20 ea

manufacturer/tradename

Swagelok® 402-1

fitting

1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

Application

Swagelok® Nut fitted with poly-dimethylsiloxane septum may be used to link stainless steel union tee for the carrier gas to enter the interface.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Aguilera-Herrador et al.
Analytical chemistry, 80(3), 793-800 (2007-12-25)
The use of ionic liquids as extracting media in single-drop liquid-phase microextraction (SDME) and its direct coupling to gas chromatography/mass spectrometry (GC/MS) is presented. For this purpose, a new removable interface that enables the introduction of the extracted analytes into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service