Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93215

Supelco

Potassium dichromate

for analysis EMPARTA® ACS

Synonym(s):

Potassium dichromate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2K2O7
CAS Number:
Molecular Weight:
294.18
MDL number:
UNSPSC Code:
12352302

grade

ACS reagent

storage temp.

20-25°C

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93215.0521: 500 g, Minimum order quantity: 1
1.93215.2521: 2.5 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service