Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21241

Supelco

GC Stationary Phase

phase OV-225, bottle of 5 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

Agency

meets requirements for USP G19

packaging

bottle of 5 g

parameter

0-265 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

OV-225 phase

solubility

acetone: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are a few parameters that should be considered in order to choose a stationary phase. OV-225 is a polar siliconised phase. It has 50% methysilicone, 25% cyanopropyl and 25% phenyl-silicone. It is possible to separate aromatic compounds from aliphatic ones containing the same number of carbons.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

OV-225 on columns is suitable for resolving neutral and amino sugars during chemical analysis using gas chromatograph.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Millar et al.
Infection and immunity, 51(1), 302-306 (1986-01-01)
Two separate species of lipopolysaccharide (LPS) from Bacteroides gingivalis 381 have been isolated. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis demonstrated not only the heterogeneity of each species, but also that they represented high- and low-molecular-weight LPS entities. Although both contained the
Michael O'Keefe
Residue Analysis in Food: Principles and Applications, 187-187 (2000)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service