Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19933

Millipore

Differential Coli-E.coli ID Membrane

white colored, sterile, diameter 70 mm, suitable for microbiology

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
$141.00

$141.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
25 DISCS
$141.00

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$141.00


Please contact Customer Service for Availability

Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

agriculture
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
Klebsiella spp.
Pseudomonas spp.
Salmonella spp.
coliforms

General description

Differential Coli-E. coli ID Membranes are substrate-carrying membranes used for rapid identification and differentiation of E. coli, Klebsiella, Pseudomonas, and Salmonella species in food and environmental samples through characteristic color reactions by placing on the agar surface post-incubation. The membranes contain chromogenic substrates such as ONPG, X-Gal, or X-Glu, and other substrates and indicators, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane. This indicates microbial enzymatic activity which serves to identify the genus and species.

Application

Differential Coli-E. coli ID Membrane is recommended for detection of E. coli, Klebsiella, Pseudomonas, and Salmonella species in the food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Escherichia coli (25922)Dark blue to violetKlebsiella pneumoniae (13883)Light pinkPseudomonas aeruginosa (27853)colorlessSalmonella Enteritidis (13076)Salmon to red

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service