Skip to Content
MilliporeSigma
All Photos(1)

Documents

20394

In-Line Pressure Gauge

pressure range 0-60 psi, gauge only (tee not included)

Synonym(s):

PRESSURE GAUGE 0-60PSIG EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

feature

gauge only (tee not included)

parameter

0-60 psi pressure range

technique(s)

gas chromatography (GC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Install a pressure gauge in the gas delivery system to measure pressure at any given point. For example, install one upstream and downstream of the High Capacity Gas Purifier. When the pressure drop across the purifier exceeds 10 psi, the converter tube should be replaced. Contains:
  • 2 in. (5 cm) diameter steel and copper alloy gauge
  • NPT to Swagelok adapter
  • 1/8 in. tee
Nuts and ferrules included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service