Key Documents
Z659290
Bandelin SONOPULS ultrasonic homogenizer accessories
Rosett cell RZ 2
Synonym(s):
Glass sample vessel
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
Pricing and availability is not currently available.
material
glass
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 3607
availability
available only in EU
volume × I.D. × H
50 mL × 42 mm × 100 mm
General description
A rosette cell (RZ) enables intensive and uniform sonication of liquid media. The round shape of the side arms leads to an intensified circulation of the sample.
Features and Benefits
  • Optimal fluid circulation .
  • Good heat dissipation .
  • Homogeneous sound reinforcement results
Choose from one of the most recent versions:
Certificates of Analysis (COA)