Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z363553

Sigma-Aldrich

Cuvette rack

Cuvette rack
1 of 1 reviewers received a sample product or took part in a promotion

Holds 20 cuvettes

Synonym(s):

rack tube, racks tube, tube racks

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$63.80

$63.80


Available to ship onApril 11, 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
1 EA
$63.80

About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

$63.80


Available to ship onApril 11, 2025Details


Request a Bulk OrderRequest more information

material

polypropylene

feature

 12.5-mm O.D. cells in two tiers

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

L × W × H

8 1/4 in. × 2 3/4 in. × 1 3/8 in.

Looking for similar products? Visit Product Comparison Guide

General description

Sloped design makes back row more accessible.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. East Lansing MI
    • Reviews 4
    • Votes 3
    5 out of 5 stars.

    Cuvette Rack

    very easy to use a clean, hope they can make a little bigger

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service