Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V3138

Sigma-Aldrich

D-Val-Leu-Lys 7-amido-4-methylcoumarin

≥98% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

mol wt

apparent mol wt 744

solubility

0.1% trifluoroacetic acid in acetonitrile: water (3:1): 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](N)C(C)C)C(=O)N[C@@H](CCCCN)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C27H41N5O5/c1-15(2)12-21(32-27(36)24(29)16(3)4)26(35)31-20(8-6-7-11-28)25(34)30-18-9-10-19-17(5)13-23(33)37-22(19)14-18/h9-10,13-16,20-21,24H,6-8,11-12,28-29H2,1-5H3,(H,30,34)(H,31,35)(H,32,36)/t20-,21-,24+/m0/s1

InChI key

NMAUGTNXBOFHCJ-AWRGLXIESA-N

Biochem/physiol Actions

Plasmin, tissue plasminogen activator substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Baker et al.
Biochimica et biophysica acta, 1117(2), 143-152 (1992-09-15)
The development of a simple, sensitive fluorimetric assay for the measurement of cell surface-associated urokinase plasminogen activator (uPA) on viable, adherent HCT116 cells in microtitre plates, after a preincubation with purified human plasminogen is described. The assay determines plasmin activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service