Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T4442

Sigma-Aldrich

Anti-Transforming Growth Factor-β2 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-TGF-β2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, pig

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

application(s)

research pathology

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGFB2(7042)

General description

Transforming Growth Factor-β 2 (TGF-β 2) is a protein produced by many cells and is found in the highest concentration in mammalian bone and porcine platelets. In body fluids like breast milk, amniotic fluid as well as the aqueous and vitreous humor of the eye, the prominent form exist is Latent TGF-β2. It has a pivotal role in regulating invasion in high-grade gliomas by inducing mesenchymal phenotype and remodeling the extracellular matrix. Anti-Transforming Growth Factor-β2 antibody can be used in neutralization and immunohistochemistry. Goat anti-TGF-β 2 antibody reacts specifically with human and porcine TGF-β 2 and TGF-β 1.2. The product has also shown less than 2% cross-reactivity for TGF-β 1, TGF-β 3 and TGF-β 5.

Specificity

The antibody recognizes TGF-β2 and TGF-β1.2. By immunoblotting (non-reducing conditions) and ELISA, the antibody shows less than 2% cross-reactivity with TGF-β1, TGF-β3, and TGF-β5.

Immunogen

porcine transforming growth factor-β2.

Application

Anti-Transforming Growth Factor-β2 antibody can be used in western blotting and ELISA.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabetta Polazzi et al.
Journal of neurochemistry, 110(2), 545-556 (2009-05-22)
Microglia, the immune cells of the CNS, play essential roles in both physiological and pathological brain states. Here we have used an in vitro model to demonstrate neuroprotection of a 48 h-microglial conditioned medium (MCM) towards cerebellar granule neurons (CGNs)
Fusun Baumann et al.
Neuro-oncology, 11(4), 368-380 (2008-11-27)
Lactate dehydrogenase type A (LDH-A) is a key metabolic enzyme catalyzing pyruvate into lactate and is excessively expressed by tumor cells. Transforming growth factor-beta2 (TGF-beta2) is a key regulator of invasion in high-grade gliomas, partially by inducing a mesenchymal phenotype
A B Roberts et al.
Molecular reproduction and development, 32(2), 91-98 (1992-06-01)
The TGF-beta's are multifunctional, pleiotropic molecules with major effects in control of cellular migration, cellular proliferation, and elaboration of extracellular matrix. Thus far, five distinct isoforms of TGF-beta have been described, each approximately 65-85% homologous and containing the characteristic 9
Lei Hong et al.
Cell death & disease, 11(8), 695-695 (2020-08-23)
Endothelial to mesenchymal transition (EndMT) is an important pathological change in many diseases. Semaphorin7A (Sema7A) has been reported to regulate nerve and vessel homeostasis, but its role in EndMT remains unclear. Here we investigate the effect of Sema7A on EndMT
Gaurav K Keshri et al.
PloS one, 11(11), e0166705-e0166705 (2016-11-20)
Chronic non-healing cutaneous wounds are often vulnerable in one or more repair phases that prevent normal healing and pose challenges to the use of conventional wound care modalities. In immunosuppressed subject, the sequential stages of healing get hampered, which may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service