MilliporeSigma
  • Lactate promotes glioma migration by TGF-beta2-dependent regulation of matrix metalloproteinase-2.
Lactate promotes glioma migration by TGF-beta2-dependent regulation of matrix metalloproteinase-2.
Neuro-oncology (2008-11-27)
Fusun Baumann, Petra Leukel, Anett Doerfelt, Christoph P Beier, Katja Dettmer, Peter J Oefner, Michael Kastenberger, Marina Kreutz, Thomas Nickl-Jockschat, Ulrich Bogdahn, Anja-Katrin Bosserhoff, Peter Hau
ABSTRACT
Lactate dehydrogenase type A (LDH-A) is a key metabolic enzyme catalyzing pyruvate into lactate and is excessively expressed by tumor cells. Transforming growth factor-beta2 (TGF-beta2) is a key regulator of invasion in high-grade gliomas, partially by inducing a mesenchymal phenotype and by remodeling the extracellular matrix. In this study, we tested the hypothesis that lactate metabolism regulates TGF-beta2-mediated migration of glioma cells. Small interfering RNA directed against LDH-A (siLDH-A) suppresses, and lactate induces, TGF-beta2 expression, suggesting that lactate metabolism is strongly associated with TGF-beta2 in glioma cells. Here we demonstrate that TGF-beta2 enhances expression, secretion, and activation of matrix metalloproteinase-2 (MMP-2) and induces the cell surface expression of integrin alpha(v)beta(3) receptors. In spheroid and Boyden chamber migration assays, inhibition of MMP-2 activity using a specific MMP-2 inhibitor and blocking of integrin alpha(v)beta(3) abrogated glioma cell migration stimulated by TGF-beta2. Furthermore, siLDH-A inhibited MMP2 activity, leading to inhibition of glioma migration. Taken together, we define an LDH-A-induced and TGF-beta2-coordinated regulatory cascade of transcriptional regulation of MMP-2 and integrin alpha(v)beta(3). This novel interaction between lactate metabolism and TGF-beta2 might constitute a crucial mechanism for glioma migration.