Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2891

Sigma-Aldrich

Tryptamine

≥99%, crystalline

Synonym(s):

2-(3-Indolyl)ethylamine, 3-(2-Aminoethyl)indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22
Beilstein:
125513
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥99%

form

crystalline

color

white

bp

137 °C/0.15 mmHg (lit.)

mp

113-116 °C (lit.)

storage temp.

−20°C

SMILES string

NCCc1c[nH]c2ccccc12

InChI

1S/C10H12N2/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6,11H2

InChI key

APJYDQYYACXCRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Vasoactive; may have a neuromodulator function; biogenic amine formed from the decarboxylation of tryptophan by L-aromatic amino acid decarboxylase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

365.0 °F - closed cup

Flash Point(C)

185 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tryptamine may couple dopaminergic and serotonergic transmission in the brain.
A V Juorio et al.
General pharmacology, 21(5), 613-616 (1990-01-01)
A J Greenshaw
Progress in neuro-psychopharmacology & biological psychiatry, 13(3-4), 431-443 (1989-01-01)
1. The relevance of trace amine research is outlined for PEA and T in the context of psychotherapeutic drug action, particularly in relation to the actions of MAO-inhibitor antidepressant drugs. 2. Evidence for the neuronal localization of these amines and
D D Mousseau
Metabolic brain disease, 8(1), 1-44 (1993-03-01)
Although early interest in the biomedical relevance of tryptamine has waned in recent years, it is clear from the above discussion that the study of tryptamine is worthy of serious consideration as a factor in neuropsychiatric disorders. The study of
6-halogenochromones bearing tryptamine: one-step access to potent and highly selective inhibitors of breast cancer resistance protein.
Glaucio Valdameri et al.
ChemMedChem, 7(7), 1177-1180 (2012-05-23)
Gabriele Loers et al.
Journal of neurochemistry, 128(1), 88-100 (2013-08-21)
Polysialic acid (PSA) is a major regulator of cell-cell interactions in the developing nervous system and in neural plasticity in the adult. As a polyanionic molecule with high water-binding capacity, PSA increases the intercellular space generating permissive conditions for cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service