Skip to Content
MilliporeSigma
Tryptamine analytical standard

Tryptamine

Tryptamine analytical standard
Synonym(s):
2-(3-Indolyl)ethylamine, 3-(2-Aminoethyl)indole
Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing