Skip to Content
MilliporeSigma
All Photos(1)

Documents

T1793

Sigma-Aldrich

Nalgene® centrifuge tubes, Oak Ridge Style 3118

nominal capacity 30 mL

Synonym(s):

High-Speed ​​Centrifuge Tubes, Nalgene tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
eCl@ss:
32119210
NACRES:
NB.11

material

polypropylene screw closure
round bottom tube
screw top cap
transparent polycarbonate

sterility

non-sterile

feature

RCF 50,000 × g

packaging

pack of 10 ea

manufacturer/tradename

Nalgene® 3118-0030

nominal capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

“Nalgene” Oak Ridge high-speed polycarbonate centrifuge tubes offer exceptional mechanical strength, making them suitable for a wide range of applications. Additionally, their transparency allows for effortless observation of the tube′s contents.

Features and Benefits

  • Can withstand temperatures ranging from -135°C to +135°C (-211°F to +275°F).
  • Designed with a round bottom to fit high-speed rotors.
  • Suitable for use in both refrigerated and non-refrigerated centrifuges.
  • Equipped with polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
  • When paired with a Nalgene sealing cap, the 30mL tubes can handle centrifugal forces up to 50,000 x g.
  • Autoclavable.

Other Notes

  • Polycarbonate centrifuge tubes will have their lifespan reduced with repeated autoclaving.
  • For proper performance, it is essential to fill the tubes to a minimum of 80% of their total capacity.
  • For leakproof performance exceeding 10,000 x g, utilize a sealing cap or sealing cap assembly.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service