Non-alkaline or neutral pH detergents are recommended for cleaning plastic labware, particularly those made of polycarbonate, due to sensitivity to alkaline attack. Please see the link below to review product 107553, an example of a neutral pH detergent:
https://www.sigmaaldrich.com/product/mm/107553
T1543
Nalgene® centrifuge tubes, Oak Ridge Style 3118
nominal capacity 10 mL
Synonym(s):
High-Speed Centrifuge Tubes, Nalgene tubes
Select a Size
$253.00
Available to ship onApril 04, 2025Details
Select a Size
About This Item
$253.00
Available to ship onApril 04, 2025Details
Recommended Products
material
polypropylene screw closure
round bottom tube
screw top cap
transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3118-0010
nominal capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Can withstand temperatures ranging from -135°C to +135°C (-211°F to +275°F).
- Designed with a round bottom to fit high-speed rotors.
- Suitable for use in both refrigerated and non-refrigerated centrifuges.
- Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
- The tubes are autoclavable.
Other Notes
- Repeated autoclaving of polycarbonate centrifuge tubes may shorten their lifespan.
- At least 80% of the total capacity must be filled to ensure proper performance.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Which is the recommended detergent for cleaning the tubes and caps?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service