Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0905

Sigma-Aldrich

Tyramine Oxidase from Arthrobacter sp.

lyophilized powder, ≥3 units/mg solid

Synonym(s):

Amine:oxygen oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥3 units/mg solid

storage temp.

2-8°C

Unit Definition

One unit will oxidize 1.0 μmole of tyramine to p-hydroxyphenylacetaldehyde per min at pH 7.5 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Narattaphol Charoenphandhu et al.
Molecular and cellular biochemistry, 375(1-2), 49-58 (2012-12-05)
Estrogen deficiency increases the risk of anxiety and mood disorders, presumably by deranging metabolism of the monoamine neurotransmitters and the expression of their reuptake transporters in the brain. Although estrogen-deficient individuals were also susceptible to stress, little was known regarding
V I Fedchenko et al.
Biomeditsinskaia khimiia, 58(5), 599-607 (2013-01-08)
Renalase is a recently discovered secretory protein, which is suggested to play a role (which still remains elusive) in regulation of blood pressure. Earlier it was purified from urine of healthy volunteers by means of ammonium sulfate fractionation and subsequent
M Bellani et al.
Epidemiology and psychiatric sciences, 21(4), 347-351 (2012-11-24)
In a short series of articles, we will review the evidence for genotype by environment interaction (G × E) in developmental psychopathology. We will focus specifically on the characteristics of types of exposure assessed with respect to both their methods and findings.
Nibha Mishra et al.
Bioorganic & medicinal chemistry letters, 23(3), 702-705 (2013-01-02)
MAO-B and AChE are the two validated targets for Alzheimer's disease. In pursuit of a single molecule hitting both the targets, we explored a set of previously reported extremely potent MAO-B selective inhibitors, for their additional AChE inhibitory activity. We
Zaixin Li et al.
Journal of medicinal food, 15(12), 1057-1063 (2012-12-12)
Memory impairment is one of main clinical symptoms of brain senescence. To address the effects of Cordyceps militaris Link extract (CE) on memory impairment, a D-galactose (D-Gal)-induced aging mouse model was employed. Mice injected with D-Gal showed a significant learning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service