Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK526

Sigma-Aldrich

Diamine Oxidase Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

detection method

fluorometric

relevant disease(s)

cancer; immunological diseases; gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Diamine oxidase (DAO) also known as histaminase or amine oxidase (copper containing), is an enzyme involved in the metabolism, oxidation, and inactivation of histamine in animals. Highest levels are observed in the digestive tract and placenta. An imbalance between histamine intake and the capacity for histamine degradation can lead to histamine intolerance (HIT). Measuring DAO activity in serum can be useful in diagnosing HIT.

Features and Benefits

  • Linear detection range 0.5 to 6 U/L .
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for diamine oxidase activity in serum or plasma samples.

Principle

The Diamine Oxidase Assay Kit is based on the oxidation of putrescine to pyrroline plus NH3 and H2O2. The generated H2O2 is then used by HRP to oxidize a dye making it fluorescent. The increase in fluorescence at λEx = 530 nm/λEm = 585 nm is directly proportional to the enzyme activity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service