Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0888

Sigma-Aldrich

Glycerol trihexanoate

~99%

Synonym(s):

1,2,3-Tricaproylglycerol, Glycerol tricapronate, Tricaproin, Trihexanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4COOCH2]2CHOCO(CH2)4CH3
CAS Number:
Molecular Weight:
386.52
Beilstein:
1806732
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

~99%

form

liquid

density

0.98 g/mL (lit.)

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCC(=O)OCC(COC(=O)CCCCC)OC(=O)CCCCC

InChI

1S/C21H38O6/c1-4-7-10-13-19(22)25-16-18(27-21(24)15-12-9-6-3)17-26-20(23)14-11-8-5-2/h18H,4-17H2,1-3H3

InChI key

MAYCICSNZYXLHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Aurousseau
The British journal of nutrition, 60(3), 525-538 (1988-11-01)
1. Results of 138 nitrogen balance studies from experiments with forty male Limousin and nineteen male Ile de France preruminant lambs were used to estimate total N requirements and to assess the effects of dietary tricaproin inclusion, protein concentration, lysine
C Lutton et al.
Annals of nutrition & metabolism, 27(2), 162-171 (1983-01-01)
The effects of a diet consisting of 10% medium-chain triglycerides (C8:0, C10:0) or 10% homogeneous triglycerides of 6- to 14-carbon chain saturated fatty acids on cholesterol turnover processes were studied in rats using the isotope equilibrium method. Cholesterol absorption was
S Fontagné et al.
The Journal of nutrition, 130(8), 2009-2015 (2000-08-05)
We investigated the effect of chain length of dietary medium-chain fatty acids (MCFA) on growth performance and fatty acid composition of first-feeding carp larvae. In a first trial, five semi-purified isolipidic (23-24 g/100 g of dry matter) diets were formulated
G J A Wanten et al.
European journal of clinical investigation, 32(4), 285-289 (2002-04-16)
Unsaturated fatty acids are known as neutrophil activators. In the present study we investigated whether saturated triglycerides and fatty acids may also contribute to the previously observed activation of neutrophils by nutritional lipid emulsions. Furthermore we tested the hypothesis that
Haibo Xu et al.
Food chemistry, 188, 439-445 (2015-06-05)
An acidic cutinase (TtcutB) from Thielavia terrestris CAU709 was purified to apparent homogeneity with 983 Um g(-1) specific activity. The molecular mass of the enzyme was estimated to be 27.3 and 27.9 kDa by SDS-PAGE and gel filtration, respectively. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service