MilliporeSigma
  • Characterization of an acidic cold-adapted cutinase from Thielavia terrestris and its application in flavor ester synthesis.
Characterization of an acidic cold-adapted cutinase from Thielavia terrestris and its application in flavor ester synthesis.
Food chemistry (2015-06-05)
Haibo Xu, Qiaojuan Yan, Xiaojie Duan, Shaoqing Yang, Zhengqiang Jiang
ABSTRACT
An acidic cutinase (TtcutB) from Thielavia terrestris CAU709 was purified to apparent homogeneity with 983 Um g(-1) specific activity. The molecular mass of the enzyme was estimated to be 27.3 and 27.9 kDa by SDS-PAGE and gel filtration, respectively. A peptide sequence homology search revealed no homologous cutinases from T. terrestris, except for one putative cutinase gene (XP003656017.1), indicating that TtcutB is a novel enzyme. TtcutB exhibited an acidic pH optimum of 4.0, and stability at pH 2.5-10.5. Optimal activity was at 55 °C, it was stable up to 65 °C, and retained over 30% activity at 0 °C. Km values toward p-nitrophenyl (pNP) acetate, pNP-butyrate and pNP-caproate were 8.3, 1.1 and 0.88 mM, respectively. The cutinase exhibited strong synthetic activity on flavor ester butyl butyrate under non-aqueous environment, and the highest esterification efficiency of 95% was observed under the optimized reaction conditions. The enzyme's unique biochemical properties suggest great potential in flavor esters-producing industries.
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
4-Nitrophenyl myristate, ≥95.0% (HPLC)
Sigma-Aldrich
4-Methylumbelliferyl butyrate, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)
Sigma-Aldrich
Triacetin, 99%, FCC, FG
Sigma-Aldrich
Triacetin, 99%
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate