Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0629

Sigma-Aldrich

L-Tryptophanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O · HCl
CAS Number:
Molecular Weight:
239.70
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white to off-white

SMILES string

Cl.N[C@@H](Cc1c[nH]c2ccccc12)C(N)=O

InChI

1S/C11H13N3O.ClH/c12-9(11(13)15)5-7-6-14-10-4-2-1-3-8(7)10;/h1-4,6,9,14H,5,12H2,(H2,13,15);1H/t9-;/m0./s1

InChI key

WOBDANBSEWOYKN-FVGYRXGTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cássia Alessandra Marquezin et al.
Biopolymers, 71(5), 569-576 (2003-11-25)
We present results of time resolved fluorescence measurements performed in Tryptophan (Trp) derivatives and Trp-containing peptides in the pH range 3.0-11.0. For each compound a set of decay profiles measured in a given range of pH values was examined as
D Andrews et al.
European journal of biochemistry, 146(1), 201-209 (1985-01-02)
When tryptophanyl-tRNA synthetase from Escherichia coli is allowed to react with L-tryptophan and ATP-Mg in the presence of inorganic pyrophosphatase, the fluorescence change of the reaction mixture reveals three or four sequential processes, depending on the buffer used. Quenched-flow and
Krishanu Ray et al.
The journal of physical chemistry. C, Nanomaterials and interfaces, 112(46), 17957-17963 (2009-01-31)
In recent years metal-enhanced fluorescence (MEF) using silver particles has been reported for a number of fluorophores emitting at visible wavelengths. However it was generally thought that silver particles would always quench fluorescence at shorter wavelengths. We now report the
Roy W Alston et al.
Biophysical journal, 94(6), 2280-2287 (2007-12-11)
This article probes the denatured state ensemble of ribonuclease Sa (RNase Sa) using fluorescence. To interpret the results obtained with RNase Sa, it is essential that we gain a better understanding of the fluorescence properties of tryptophan (Trp) in peptides.
D Pörschke et al.
Nucleic acids research, 10(19), 6163-6176 (1982-10-11)
The amides of L-phenylalanine, L-tyrosine and L-tryptophane decrease the melting temperatures tm of poly(A)*poly(U) and poly(I)*poly(C) double helices at low concentrations (1 mM), whereas high concentrations finally lead to an increase of tm. This dependence of the tm-values upon the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service