Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

604836

Sigma-Aldrich

L-Tryptophan-1-13C

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC10H12N2O2
CAS Number:
Molecular Weight:
205.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

optical activity

[α]20/D -30.5°, c = 1 in H2O

mp

280-285 °C (dec.) (lit.)

mass shift

M+1

SMILES string

N[C@@H](Cc1c[nH]c2ccccc12)[13C](O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i11+1

InChI key

QIVBCDIJIAJPQS-KYURPWGOSA-N

General description

L-Tryptophan-1-13C is an isotopic analogue of L-tryptophan.

Application

L-Tryptophan-1-13C is used in quantitative determination of proteins using mass spectrometry.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerald W Becker
Briefings in functional genomics & proteomics, 7(5), 371-382 (2008-12-25)
Straightforward methods for the introduction of stable isotopes into proteins with subsequent isolation and purification of the proteins will greatly aid the field of quantitative proteomics. Proteins containing amino acids with one or more of the stable isotopes of deuterium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service