Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5081

Sigma-Aldrich

PTPRM (764-1452), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPRM(5797)

General description

PTPRM is a member of the protein tyrosine phosphatase family and can participate in a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTPRM has been shown to mediate cell-cell aggregation through the interaction with another molecule of PTPRM on an adjacent cell. PTPRM can interact with scaffolding protein RACK1/GNB2L1 and this interaction may be necessary for downstream signaling in response to cell-cell adhesion. PTPRM has been shown to be expressed in human pulmonary vascular endothelia where it directly binds to VE-cadherin and regulates both the tyrosine phosphorylation state of VE-cadherin and barrier integrity.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elianne A Koop et al.
The International journal of developmental biology, 47(5), 345-354 (2003-08-05)
The receptor-like protein tyrosine phosphatase mu (RPTPmu) belongs to the subfamily of meprin, A5, RPTPmu (MAM) domain-containing RPTPs, which are thought to play an important role in cell-cell adhesion mediated processes. The current study was designed to examine the expression
Polly J Phillips-Mason et al.
The Journal of biological chemistry, 281(8), 4903-4910 (2005-12-29)
The receptor protein-tyrosine phosphatase PTPmu is a member of the Ig superfamily of cell adhesion molecules. The extracellular domain of PTPmu contains motifs commonly found in cell adhesion molecules. The intracellular domain of PTPmu contains two conserved catalytic domains, only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service