MilliporeSigma
  • Receptor protein tyrosine phosphatase mu expression as a marker for endothelial cell heterogeneity; analysis of RPTPmu gene expression using LacZ knock-in mice.
Receptor protein tyrosine phosphatase mu expression as a marker for endothelial cell heterogeneity; analysis of RPTPmu gene expression using LacZ knock-in mice.
The International journal of developmental biology (2003-08-05)
Elianne A Koop, Susana M Chuva de Sousa Lopes, Elles Feiken, Hans A R Bluyssen, Martin van der Valk, Emile E Voest, Christine L Mummery, Wouter H Moolenaar, Martijn F B G Gebbink
ABSTRACT
The receptor-like protein tyrosine phosphatase mu (RPTPmu) belongs to the subfamily of meprin, A5, RPTPmu (MAM) domain-containing RPTPs, which are thought to play an important role in cell-cell adhesion mediated processes. The current study was designed to examine the expression pattern of RPTPmu in mice. We have generated RPTPmu-LacZ knock-in mice that express the beta-galactosidase (LacZ) reporter gene under the control of the RPTPmu promoter. LacZ expression patterns were analysed in embryos and adult mice by whole mount LacZ staining. Analysis of beta-galactosidase activity of heterozygous embryos and adult tissues revealed RPTPmu expression in endothelial cells of arteries and capillaries. In contrast, expression was virtually absent in endothelial cells of veins and in fenestrated endothelial cells in the adult liver and spleen. Moreover, RPTPmu expression was found in endothelial cells from the endocardium and the aorta in embryos, but not in adult mice. In addition to heterogeneous expression in endothelial cells, RPTPmu expression was found in cardiac muscle cells but not in skeletal muscle cells or smooth muscle cells. Expression was also found in Type II pneumonocytes in the lung alveoli and in Purkinje cells and other neurons in the brain. The specific expression of RPTPmu in arterial endothelial cells and in cardiac myocytes suggests that RPTPmu may play a role in the regulation of cardiovascular functions.