Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5021

Sigma-Aldrich

DUSP22, active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JKAP, JSP1, VHX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

510-690 nmol/min·mg

mol wt

~43 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DUSP22(56940)

General description

Dual specificity phosphatases (DUSP) family, which can dephosphorylate both phosphoserine/threonine and phosphotyrosine residues, comprise of two subgroups: "typical" DUSP (include 11 MKPs that dephosphorylate the mitogen-activated protein kinases Erk, Jnk, and p38 at their dually phosphorylated TXY activation loop motif) and "atypical" DUSP (include 8 enzymes that lack specific MAP kinase-targeting motifs and tend to be much smaller proteins, which are less than 250 amino acid residues). DUSP22, also called JKAP, JSP1 and MKPX plays a role in various inflammatory and proliferative disorders associated with dysfunctional Jnk signaling.

Physical form

Supplied in 20mM MOPS, pH 7.5, 50mM NaCl, 10mM glutathione, 0.25mM DTT, 0.1mM PMSF, 30% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andres Alonso et al.
The Journal of biological chemistry, 279(31), 32586-32591 (2004-05-13)
The human DUSP15 gene encodes an uncharacterized 235-amino acid member of the subfamily of small dual specificity protein phosphatases related to the Vaccinia virus VH1 phosphatase. Similar to VHR-related MKPX (VHX) (DUSP22), the predicted protein has an N-terminal myristoylation recognition
Y Shen et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(24), 13613-13618 (2001-11-22)
The mitogen-activated protein kinases (MAPKs) are integral to the mechanisms by which cells respond to physiological stimuli, such as growth factors, hormones, and cytokines, and to a wide variety of environmental stresses. The MAPKs, which are stimulated by phosphorylation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service