MilliporeSigma
  • VHY, a novel myristoylated testis-restricted dual specificity protein phosphatase related to VHX.
VHY, a novel myristoylated testis-restricted dual specificity protein phosphatase related to VHX.
The Journal of biological chemistry (2004-05-13)
Andres Alonso, Sonoko Narisawa, Jori Bogetz, Lutz Tautz, Radinka Hadzic, Huong Huynh, Scott Williams, Anette Gjörloff-Wingren, Meire C D Bremer, Leslie J Holsinger, José L Millan, Tomas Mustelin
ABSTRACT
The human DUSP15 gene encodes an uncharacterized 235-amino acid member of the subfamily of small dual specificity protein phosphatases related to the Vaccinia virus VH1 phosphatase. Similar to VHR-related MKPX (VHX) (DUSP22), the predicted protein has an N-terminal myristoylation recognition sequence, and we show here that both are indeed modified by the attachment of a myristate to Gly-2. In recognition of this relatedness to VHX, we refer to the DUSP15-encoded protein as VH1-related member Y (VHY). We report that VHY is expressed at high levels in the testis and barely detectable levels in the brain, spinal cord, and thyroid. A VHY-specific antiserum detected a protein with an apparent molecular mass of 26 kDa, and histochemical analysis showed that VHY was readily detectable in pachytene spermatocytes (midstage of meiotic division I) and round spermatids and weakly in Leydig cells (somatic cells outside of the seminiferous tubules). When expressed in 293T or NIH-3T3 cells, VHY was concentrated at the plasma membrane with some staining of vesicular structures in the Golgi region. Mutation of the myristoylation site Gly-2 abrogated membrane location. Finally, we demonstrate that VHY is an active phosphatase in vitro. We conclude that VHY is a new member of a subgroup of myristoylated VH1-like small dual specificity phosphatases.
Sigma-Aldrich
DUSP22, active, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution