Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2070

Sigma-Aldrich

LRH-1 human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$589.00

$589.00


Please contact Customer Service for Availability


Select a Size

Change View
5 μG
$589.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$589.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~62.6 kDa

packaging

pkg of 5 μg

concentration

50 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NR5A2(2494)

Biochem/physiol Actions

LRH-1 or HB1F for human B1-binding factor belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors and is closely related to steroidogenic factor-1. LRH1 contains a DNA-binding domain with 2 zinc finger motifs, an FTZ-F1 box, and a ligand-binding domain. Cholesterol 7-α-hydroxylase is the first and rate-limiting enzyme in a pathway through which cholesterol is metabolized to bile acids. Elevated promoter-specific repressor protein (SHP) inactivates LRH1 by forming a heterodimeric complex that leads to promoter-specific repression of both CYP7A1 and SHP. These results revealed an elaborate autoregulatory cascade mediated by nuclear receptors for the maintenance of hepatic cholesterol catabolism. LRH1 specifically binds and activates viral hepatitis B enhancer II, an essential element for the liver-specific regulation of hepatitis B virus gene expression. CPF is a key regulator of human CYP7A gene expression in the liver. SHP1 represses expression of CYP7A1 by inhibiting the activity of LRH1 which regulates CYP7A1 expression positively. This bile acid-activated regulatory cascade provides a molecular basis for the coordinate suppression of CYP7A1 and other genes involved in bile acid biosynthesis.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu, T. T., et al.
Molecular and Cellular Biology, 6, 507-515 (2000)
Goodwin, B., et al.
Molecular and Cellular Biology, 25, 517-526 (2000)
M Li et al.
The Journal of biological chemistry, 273(44), 29022-29031 (1998-10-24)
Enhancer II (ENII) of hepatitis B virus (HBV) is one of the essential cis-elements for the transcriptional regulation of HBV gene expression. Its function is highly liver-specific, suggesting that liver-enriched transcriptional factors play critical roles in regulating the activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service