Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

676496

Sigma-Aldrich

VEGF Inhibitor, CBO-P11

The VEGF Inhibitor, CBO-P11 controls the biological activity of VEGF. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGF Inhibitor, CBO-P11, Cyclo-VEGI, cyclic( D-F-PQIMRIKPHQGQHIGE), D-Phe-Pro(79-93), VEGFR Tyrosine Kinase Inhibitor XX, VEGFR2 Kinase Inhibitor XVI, Cyclo-VEGI, cyclic(D-F-PQIMRIKPHQGQHIGE), D-Phe-Pro(79-93), VEGFR Tyrosine Kinase Inhibitor XX, VEGFR2 Kinase Inhibitor XVI

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$418.00

$418.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
$418.00

About This Item

Empirical Formula (Hill Notation):
C90H140N28O22S
Molecular Weight:
1998.31
UNSPSC Code:
12352200
NACRES:
NA.77

$418.00


Please contact Customer Service for Availability

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 5 mg/mL
water: soluble

shipped in

wet ice

storage temp.

−20°C

General description

A macrocyclic 17-amino acid peptide derived from residues 79 - 93 of vascular endothelial growth factor that mediate its binding to VEGFR-2. It blocks the binding of VEGF165 to its receptors (IC50 = 700 nM for VEGFR1, 1.3 µM for VEGFR2) and exhibits anti-angiogenic activity as well as other VEGFR-mediated cellular functions, such as proliferation (IC50 = 5.8 µM), migration (IC50 = 8.2 µM), and activation of MAP kinases. Its anti-angiogenic activity has been successfully demonstrated in vivo using chick embryos and nude mice tumor models of human intracranial and syngeneic glioma.
A macrocyclic, 17-amino acid peptide derived from residues 79-93 of vascular endothelial growth factor (VEGF) that mediates the binding of VEGF to its receptor, VEGFR-2. Blocks the binding of VEGF165 to its receptors (IC50 = 70 nM for VEGFR-1, 1.3 µM for VEGFR-2) and exhibits anti-angiogenic properties. Also reported to block other VEGF-mediated cellular functions such as proliferation (IC50 = 5.8 µM), migration (IC50 = 8.2 µM), and activation of MAP kinases. Also reported to exhibit anti-angiogenic properties in vivo using chick embryo and nude mouse tumor models of human intracranial and syngeneic glioma.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Blocks the binding of VEGF165 to its receptors
Product does not compete with ATP.
Reversible: no
Target IC50: 700 nM for VEGFR1, 1.3 µM for VEGFR2

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Sequence

cyclic(D-Phe-Pro-Gln-Ile-Met-Arg-Ile-Lys-Pro-His-Gln-Gly-Gln-His-Ile-Gly-Glu)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zilberberg, L., et al. 2003. J. Biol. Chem.278, 35564.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service