Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4078

MitoBloCK-10

new

≥98% (HPLC)

Synonym(s):

3-​Fluoro-​benzoic acid 2-​[(5-​nitro-​2-​thienyl)​methylene]​hydrazide, 3-Fluoro-N’- [(5-nitro-2-thienyl)methylene]benzohydrazide, MB-10, Mitochondrial import blockers of the Carla Koehler laboratory-10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8FN3O3S
CAS Number:
Molecular Weight:
293.27
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1cc(ccc1)C(=O)N\N=C\c2[s]c(cc2)[N+](=O)[O-]

InChI

1S/C12H8FN3O3S/c13-9-3-1-2-8(6-9)12(17)15-14-7-10-4-5-11(20-10)16(18)19/h1-7H,(H,15,17)/b14-7+

InChI key

QDYQZMWFIYLMNX-VGOFMYFVSA-N

Biochem/physiol Actions

Mitochondrial import blocker of substances requiring inner membrane translocon TIM23.
MitoBloCK-10 (MB-10) is mitochondrial import blocker of substances requiring inner membrane translocon TIM23 (TIM23) in yeast and mammalian mitochondria. It appears that MB-10 targets Tim44 of the PAM complex. MitoBloCK-10 does not alter respiraton. MB-10 inhibits cardiac development and induces apoptosis in Zebrafish embryos. It inhibits bladder cancer cell growth in vitro and in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A first-in-class TIMM44 blocker inhibits bladder cancer cell growth
Cell Death & Disease, 15(3), 204-204 (2024)
Non Miyata et al.
The Journal of biological chemistry, 292(13), 5429-5442 (2017-02-09)
Diverse protein import pathways into mitochondria use translocons on the outer membrane (TOM) and inner membrane (TIM). We adapted a genetic screen, based on Ura3 mistargeting from mitochondria to the cytosol, to identify small molecules that attenuated protein import. Small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service