MilliporeSigma
  • Adaptation of a Genetic Screen Reveals an Inhibitor for Mitochondrial Protein Import Component Tim44.
Adaptation of a Genetic Screen Reveals an Inhibitor for Mitochondrial Protein Import Component Tim44.
The Journal of biological chemistry (2017-02-09)
Non Miyata, Zhiye Tang, Michael A Conti, Meghan E Johnson, Colin J Douglas, Samuel A Hasson, Robert Damoiseaux, Chia-En A Chang, Carla M Koehler
ABSTRACT
Diverse protein import pathways into mitochondria use translocons on the outer membrane (TOM) and inner membrane (TIM). We adapted a genetic screen, based on Ura3 mistargeting from mitochondria to the cytosol, to identify small molecules that attenuated protein import. Small molecule mitochondrial import blockers of the Carla Koehler laboratory (MB)-10 inhibited import of substrates that require the TIM23 translocon. Mutational analysis coupled with molecular docking and molecular dynamics modeling revealed that MB-10 binds to a specific pocket in the C-terminal domain of Tim44 of the protein-associated motor (PAM) complex. This region was proposed to anchor Tim44 to the membrane, but biochemical studies with MB-10 show that this region is required for binding to the translocating precursor and binding to mtHsp70 in low ATP conditions. This study also supports a direct role for the PAM complex in the import of substrates that are laterally sorted to the inner membrane, as well as the mitochondrial matrix. Thus, MB-10 is the first small molecule modulator to attenuate PAM complex activity, likely through binding to the C-terminal region of Tim44.
Product Description
MitoBloCK-10, ≥98% (HPLC)