Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3845

Sigma-Aldrich

Enterocin

≥95% (HPLC)

Synonym(s):

(3R,4aR,5S,6S,7S,7aS,8R)-7-Benzoylhexahydro-4a,6,7a,8-tetrahydroxy-5-(4-methoxy-2-oxo-2H-pyran-6-yl)-3,6-methanocyclopenta[c]pyran-1(3H)-one, Vulgamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O10
CAS Number:
Molecular Weight:
444.39
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

(Powder or lyophilized powder)

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Enterocin is a bacteriostatic polyketide antibiotic unrelated to the polypeptide enterocin bacteriocins. It has broad-spectrum antibiotic activity against both gram-positive and gram-negative bacteria. Enterocin has also been shown to have inhibitory activity against β-amyloid protein (Aβ1-42) fibrillation and moderate cytotoxicity against HeLa and HepG2 cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring Natural Product Artifacts: The Polyketide Enterocin Warms to a Ballet of Isomers
Organic Letters, 22(12), 4828-4832 (2020)
Direct capture and heterologous expression of Salinispora natural product genes for the biosynthesis of enterocin
Journal of Natural Products, 78(3), 539-542 (2015)
Dong-Bo Xu et al.
Applied microbiology and biotechnology, 99(14), 5825-5832 (2015-04-22)
The type II polyketide synthase (PKS) natural product enterocin (1) was isolated from a mangrove-derived novel species Streptomyces qinglanensis 172205 guided by genome sequence, and its putative biosynthetic gene cluster was revealed. Its natural analogues 5-deoxyenterocin (2) and wailupemycin A-C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service