Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3836

Sigma-Aldrich

Delamanid

≥98% (HPLC)

Synonym(s):

(2R)-2,3-Dihydro-2-methyl-6-nitro-2-{4-[4-[4-(trifluoromethoxy)phenoxy]-1-piperidinyl]phenoxy]methyl]-imidazo[2,1-b]oxazole, (R)-2-Methyl-6-nitro-2-{4-[4-(4-trifluoromethoxyphenoxy)piperidin-1-yl]phenoxymethyl}-2,3-dihydroimidazo[2,1-b]oxazole, Deltyba, OPC 67683, OPC-67683, OPC67683

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$136.00
50 MG
$546.00

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$136.00
50 MG
$546.00

About This Item

Empirical Formula (Hill Notation):
C25H25F3N4O6
CAS Number:
Molecular Weight:
534.48
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

$136.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -8 to -12°, c = 0.5 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC(F)(F)Oc1ccc(cc1)OC2CCN(CC2)c3ccc(cc3)OC[C@@]4(Oc5[n](cc(n5)[N+](=O)[O-])C4)C

InChI

1S/C25H25F3N4O6/c1-24(15-31-14-22(32(33)34)29-23(31)38-24)16-35-18-4-2-17(3-5-18)30-12-10-20(11-13-30)36-19-6-8-21(9-7-19)37-25(26,27)28/h2-9,14,20H,10-13,15-16H2,1H3/t24-/m1/s1

InChI key

XDAOLTSRNUSPPH-XMMPIXPASA-N

Biochem/physiol Actions

Delamanid (OPC-67683) is an orally active mycolic acid biosynthesis inhibitor that is effective against both drug-susceptible and drug-resistant strains of M. tuberculosis H37Rv in vitro (MIC = 6 ng/mL) and exhibits significant in vivo efficacy with ~15-fold higher potency than rifampicin (RFP) in mice infected with M. tuberculosis Kurono (fold of lung CFU reduction post 28 days = 41.5 with 5 mg RFP/kg or 0.313 mg delamanid/kg daily orally dosage, 11,000-fold reduction with 0.625 mg delamanid/kg daily orally dosage).
Orally active mycolic acid biosynthesis inhibitor that is effective against drug-resistant strains of M. tuberculosis and is superior to rifampicin in vivo.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delamanid Kills Dormant Mycobacteria In Vitro and in a Guinea Pig Model of Tuberculosis
Antimicrobial Agents and Chemotherapy, 61(6), e02402-e02416 (2017)
Hirofumi Sasaki et al.
Journal of medicinal chemistry, 49(26), 7854-7860 (2006-12-22)
In an effort to develop potent new antituberculosis agents that would be effective against both drug-susceptible and drug-resistant strains of Mycobacterium tuberculosis, we prepared a novel series of optically active 6-nitro-2,3-dihydroimidazo[2,1-b]oxazoles substituted at the 2-position with various phenoxymethyl groups and
Makoto Matsumoto et al.
PLoS medicine, 3(11), e466-e466 (2006-11-30)
Tuberculosis (TB) is still a leading cause of death worldwide. Almost a third of the world's population is infected with TB bacilli, and each year approximately 8 million people develop active TB and 2 million die as a result. Today's

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service