Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3649

Sigma-Aldrich

Ozenoxacin

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-8-methyl-7-(5-methyl-6-methylamino-pyridin-3-yl)-4-oxo1,4-dihydro-quinoline-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O3
CAS Number:
Molecular Weight:
363.41
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Ozenoxacin is a quinolone antimicrobial that inhibits bacterial DNA replication enzymes, DNA gyrase A and topoisomerase IV

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y López et al.
Antimicrobial agents and chemotherapy, 57(12), 6389-6392 (2013-10-02)
In vitro activity of ozenoxacin, a novel nonfluorinated topical (L. D. Saravolatz and J. Leggett, Clin. Infect. Dis. 37:1210-1215, 2003) quinolone, was compared with the activities of other quinolones against well-characterized quinolone-susceptible and quinolone-resistant Gram-positive bacteria. Ozenoxacin was 3-fold to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service