MilliporeSigma
  • In vitro activity of Ozenoxacin against quinolone-susceptible and quinolone-resistant gram-positive bacteria.
In vitro activity of Ozenoxacin against quinolone-susceptible and quinolone-resistant gram-positive bacteria.
Antimicrobial agents and chemotherapy (2013-10-02)
Y López, M Tato, P Espinal, F Garcia-Alonso, D Gargallo-Viola, R Cantón, J Vila
ABSTRACT
In vitro activity of ozenoxacin, a novel nonfluorinated topical (L. D. Saravolatz and J. Leggett, Clin. Infect. Dis. 37:1210-1215, 2003) quinolone, was compared with the activities of other quinolones against well-characterized quinolone-susceptible and quinolone-resistant Gram-positive bacteria. Ozenoxacin was 3-fold to 321-fold more active than other quinolones. Ozenoxacin could represent a first-in-class nonfluorinated quinolone for the topical treatment of a broad range of dermatological infections.
Sigma-Aldrich
Ozenoxacin, ≥98% (HPLC)