Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3447

Sigma-Aldrich

GO289

≥98% (HPLC)

Synonym(s):

(E)-5-Bromo-2-methoxy-4-((3-(methylthio)-5-phenyl-4H-1,2,4-triazol-4-ylimino)methyl)phenol, 5-Bromo-2-methoxy-4-[[[3-(methylthio)-5-phenyl-4H-1,2,4-triazol-4-yl]imino]methyl]phenol, GO 289, GO-289

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrN4O2S
CAS Number:
Molecular Weight:
419.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

GO289 is a highly potent and selective casein kinase CK2 (CKII) inhibitor (IC50 = 7 nM/CK2, 13 μM/PIM2, >50 μM/CLK2; <50% inhibition of CK1α/δ/γ and 56 other kinases at 5 μM) that targets CK2-specific residues (CK2α V66, M163, and H160) with more overall interactions at the CK2 ATP-binding pocket than TBB, DMAT & CX-4945. GO289 selectively inhibits cellular CK1 substrates phosphorylation (0.8-8 μM; HEK293T, U2OS), including clock proteins PER2, CRY1, CLOCK & BMAL1, and displays circadian period-lengthening (Bmal1-dLuc USOS EC50 = 3.3 μM) as well as growth inhibitory potency in CK2-dependent human renal cell carcinoma (RCC) cultures.
Potent and highly selective CK2 (CKII) inhibitor with circadian period-lengthening and cancer growth inhibitory efficacy in human renal cell carcinoma cultures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service