Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3143

Sigma-Aldrich

CIGB-300 trifluoroacetate

≥95% (HPLC)

Synonym(s):

CIGB-325 trifluoroacetate, H-GRKKRRQRRRPPQ-βA-CWMSPRHLGTC-NH2 trifluoroacetate (βA = β-alanine; C15-C25 disulfide), H-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Pro-Pro-Gln-βAla-Cys-Trp-Met-Ser-Pro-Arg-His-Leu-Gly-Thr-Cys-NH2 trifluoroacetate (Cys15-Cys25 disulfide), P15-Tat trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C127H216N54O29S3
CAS Number:
Molecular Weight:
3059.61
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

film

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

CIGB-300 (P15-Tat) is a substrate-competitive casein kinase 2 inhibitor (CK2, CKII) composed of a membrane-permeant HIV-1 Tat protein transduction domain sequence (a.a. 48-60) N-terminal to a disulfide-bridged cyclic substrate peptide that inhibits against CK2-mediated substrate HPV-16 E7 phosphorylation in vitro. CIGB-300 exhibits cytotoxic effect in various cancer cultures (IC50 from 20 to 182 μM) via apoptosis induction and effectively inhibits TC-1 tumor growth in mice in vivo (48 μmol/200 μL/mouse via daily intratumoral injection).
Substrate-competitive casein kinase 2 (CK2, CKII) inhibitor with anti-cancer efficacy in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service