Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3281

Sigma-Aldrich

GSK215 hydrochloride

≥98% (HPLC)

Synonym(s):

(2S,4R)-4-hydroxy-1-((S)-2-(2-(4-(3-methoxy-4-(4-(2-(methylcarbamoyl)phenylamino)-5-(trifluoromethyl)pyridin-2-ylamino)phenyl)piperazin-1-yl)acetamido)-3,3-dimethylbutanoyl)-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride, GSK 215 hydrochloride, GSK-215 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00
25 MG
$711.00

$177.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$177.00
25 MG
$711.00

About This Item

Empirical Formula (Hill Notation):
C50H59F3N10O6S · xHCl
Molecular Weight:
985.13 (free base basis)
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

$177.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

composition

, 0-8 (H2O)
, 1.8-3.3 mol (HCl)

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cnc(cc1Nc7c(cccc7)C(=O)NC)Nc2c(cc(cc2)N3CCN(CC3)CC(=O)N[C@@H](C(C)(C)C)C(=O)N4[C@@H](C[C@H](C4)O)C(=O)N[C@@H](C)c5ccc(cc5)c6[s]cnc6C)OC

InChI key

ZGSWGXNEXAXEGV-XFCHVEHOSA-N

Biochem/physiol Actions

GSK215 is a potent and selective focal adhesion kinase (FAK) degrader (DC50 = 1.3 nM; A549 cells) composed of the FAK inhibitor VS-4718 and the VHL E3 ligase binder VHL-021. GSK215 shows cell type-dependent growth inhibition, where it inhibits A549 (IC50 <1 µM) and MCF-7 (IC50 <0.1 µM), but not BT474, in 2D cultures, while the growth of BT47 (IC50 <0.1 µM) and MCF-7 (IC50 ~0.1 µM), but not A5494, are inhibited by GSK215 in 3D cultures.
Potent and selective focal adhesion kinase (FAK) degrader composed of the FAK inhibitor VS-4718 and the VHL E3 ligase binder VHL-021.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and Characterisation of Highly Cooperative FAK-Degrading PROTACs
Angewandte Chemie (International Edition in English), 18;60(43), 23327-23334 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service