Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2713

Sigma-Aldrich

MU1210

≥98% (HPLC)

Synonym(s):

1-Methyl-4-(9-(3-(pyridin-4-yl)-phenyl)-7-oxa-2-aza-bicyclo[4.3.0]nona-1,3,5,8-tetraen-3-yl)-1H-pyrazole, 5-(1-Methyl-1H-pyrazol-4-yl)-3-(3-(pyridin-4-yl)phenyl)furo[3,2-b]pyridine, VN339

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16N4O
CAS Number:
Molecular Weight:
352.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(N=C1)C=C1C2=CC=C3C(C(C4=CC=CC(C5=CC=NC=C5)=C4)=CO3)=N2

Biochem/physiol Actions

MU1210 is a potent and selective inhibitor against cdc-like kinase (cdc2-like kinase) CLK1, CLK2, CLK4 (CLK1/2/4 IC50 = 8/20/12 nM; CLK1/2/4 Ki = 84/91/23 nM by NanoBRET) and HIPK2 (IC50 = 29 nM) with good selectivity over HIPK1/3 (IC50 = 187/159 nM), DYRK1a/1b/2 (IC50 = 213/956/1309 nM), CLK3 (IC50 >3 μM) and 194 other kinases. MU1210 enhances alternative spliced MDM4-S mRNA (10 μM, 24 h) and exhibits antiproliferation activity in MCF7 cultures (IC50 in 72 h = 1.1 μM). Unlike most ATP site-targeting kinase inhibitors, X-ray crystallography shows that MU1210 is anchored to the back pocket instead of the hinge region of CLK1. MU140 serves as a negative control.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Václav Němec et al.
Angewandte Chemie (International ed. in English), 58(4), 1062-1066 (2018-12-21)
Reported is the identification of the furo[3,2-b]pyridine core as a novel scaffold for potent and highly selective inhibitors of cdc-like kinases (CLKs) and efficient modulators of the Hedgehog signaling pathway. Initially, a diverse target compound set was prepared by synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service